Anti-CSRP1 Rabbit Polyclonal Antibody

Supplier: Genetex
GTX53932
10792-378EA 623.1 USD
10792-378
Anti-CSRP1 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to CSRP1

Type: Primary
Antigen: CSRP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rabbit
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR