Anti-TCL1A Rabbit Polyclonal Antibody

Supplier: Genetex
GTX16378
10792-120EA 623.1 USD
10792-120
Anti-TCL1A Rabbit Polyclonal Antibody
Antibodies
Rabbit Polyclonal antibody to TCL1A

Type: Primary
Antigen: TCL1A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR