Lecithin, high purity

Supplier: VWR

0579-250G 0579-1KG
10791-820CS 878.59 USD
10791-820 10791-822
Lecithin, high purity
Lecithin
Density: 1.0305 g/cm³ (20 °C)
MDL Number: MFCD00147406
CAS Number: 8002-43-5

Order Now

Specification Test Results

Acetone Insolubles = 50.0 %
Acid Value = 36
Hexane Insolubles = 0.3 %
Moisture (Karl Fischer) = 1.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR