Cannon Bath Cooler, 230 V, Cannon Instrument Company
Supplier: Cannon
9726-D10
10791-432EA 7146.73 USD
10791-432
Cannon Bath Cooler, 230 V, Cannon Instrument Company
Circulating Thermostats Closed Circulator Systems Cooling Circulators
The compact and efficient Cannon Bath Cooler (CBC) uses an advanced technological design to maintain a liquid bath below room temperature
With the CBC, bath temperatures can be lowered up to 15°C below ambient. The CBC consists of a thermoelectric Cooling Unit and a separate power supply.