HardyDisks™ AST Ceftazidime, CAZ-30, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS
Z8271
10790-418EA
33.87
USD
10790-418
HardyDisks™ AST Ceftazidime, CAZ-30, Hardy Diagnostics
Microbiology Test Systems
Antibiotic Tests
Ceftazidime, CAZ-30, HardyDisks™ AST, for susceptibility testing.
HardyDisk™ AST’s are impregnated paper disks used for Antimicrobial Susceptibility Testing (AST); also known as disk diffusion or Kirby-Bauer testing.
Each cartridge contains 50 disks that are spring loaded for smooth and reliable dispensing. The HardyDisk™ AST cartridges are designed to be fully compatible with the BBL single or multiple disk dispensers.
Delivery information: Freeze upon arrival.
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)