Lugol's iodine solution, Dobell and O'Connor stain, used in parasitology
Supplier: HARDY DIAGNOSTICS
Warning
Z66
10790-370EA 74.37 USD
10790-370
Lugol's iodine solution, Dobell and O'Connor stain, used in parasitology
Lugol's iodine solution
For use in staining parasite ova and protozoan cysts in fecal wet mounts.
This preparation is a 1:5 dilution of Lugol's Iodine.
The Hardy Diagnostics manufacturing facility and quality management system comply with the FDA's Good Manufacturing Practices (GMP) and have achieved ISO 13485 certification as a medical device manufacturer.