Transport Swabs, Liquid Amies, Dual Swab
Supplier: Hardy Diagnostics
4138BX
10790-364EA 111.96 USD
10790-364
Transport Swabs, Liquid Amies, Dual Swab
Swabs Transport Swabs
For throat, vaginal, wound and skin swabs.
  • Tamper-evident seal to assure first time use
  • Double plastic shaft
  • Regular rayon tip
Sterile soft rayon swabs on two plastic shafts with a round bottom plastic tube containing liquid modified Amies transport medium for general microbiology and plate count purposes, contains no agar. Packaged in a clear oxygen barrier wrapper, keeps out oxygen, tamper-evident seal.
Amies media is Stuart's original media formula modified by replacing the glycerophosphate in the original formula for an inorganic phosphate buffer. The metabolism of glycerophosphate by coliform organisms and other Gram-negative rods, in Stuart's original formulation, resulted in proliferation of these organisms from throat, wound and fecal specimens. Amies also found that NaCl at 0.3% w/v was optimal for the preservation of Neisseria gonorrhoeae. Calcium and magnesium salts were added in the belief that these ions were of importance in controlling the permeability of the bacterial cells and contributed to their survival.