LanaGram™ Rapid Color Test, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS
Z49
10789-718PK 114.51 USD
10789-718
LanaGram™ Rapid Color Test, Hardy Diagnostics
Stains Microbiology Stains
LanaGram™ rapid color test for bacterial gram reaction.
For organisms that are difficult to gram stain
LanaGram™ produces a rapid color change in a tube, gram negatives turn yellow, gram positives remain clear. Ideal for Bacillus, Listeria, Lactobacillus, and non-fermentors, all of which can be difficult to gram stain.
The Hardy Diagnostics manufacturing facility and quality management system comply with the FDA's Good Manufacturing Practices (GMP) and have achieved ISO 13485 certification as a medical device manufacturer.
Uses the aminopeptidase reaction. LanaGram™ confirms the gram stain reaction of gram-negative and gram-positive aerobic or facultatively anaerobic rods or coccobacilli. This test is not recommended for use with gram-positive cocci, microaerophilic or obligate anaerobic organisms.