HardyDisks™ AST Cefazolin, CZ-30, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS
Z8145
10789-438EA 90.56 USD
10789-438
HardyDisks™ AST Cefazolin, CZ-30, Hardy Diagnostics
Microbiology Test Systems Antibiotic Tests
Cefazolin, CZ-30, HardyDisks™ AST, for susceptibility testing.
HardyDisk™ AST’s are impregnated paper disks used for Antimicrobial Susceptibility Testing (AST); also known as disk diffusion or Kirby-Bauer testing.
Each cartridge contains 50 disks that are spring loaded for smooth and reliable dispensing. The HardyDisk™ AST cartridges are designed to be fully compatible with the BBL single or multiple disk dispensers. The last disk in each cartridge is clearly marked so as to alert you when refills are needed.
Delivery information: Freeze upon arrival.