Miriad Rapid HBc/HIV/HCV Antibody Test POU+, MedMira
Supplier: MEDMIRA
815311006005
10789-423CS 11478.75 USD
10789-423
Miriad Rapid HBc/HIV/HCV Antibody Test POU+, MedMira
Clinical Diagnostic Supplies Clinical Reagents and Kits
Miriad Rapid HBc/HIV/HCV Antibody Test POU+ employs MedMira rapid vertical flow technology to deliver the simultaneous diagnosis of hepatitis B core, HIV-1/2 and hepatitis C
Miriad Rapid HBc/HIV/HCV Antibody Test POU+ features a built-in reagent control line, works with whole blood, serum and plasma specimens, and requires no additional equipment. Test kits can be stored for up to 18 months at 2–30°C (35.6–86°F).
In three steps, users can obtain multiple instant results on a single test cartridge.
Ordering information: HBc/HIV/HCV Antibody Test Controls are available as an accessory to the test (10769-911).
Delivery information: Each box of Miriad Rapid HBc/HIV/HCV Test POU+ includes 25 test kits with each test kit containing 1 test cartridge, 1 InstantGold cap, 1 Buffer Vial 1, 1 Buffer Vial 2, 1 transfer pipette and 1 package insert.