CompactDry™ LS, Listeria spp, Hardy Diagnostics
Supplier: HARDY DIAGNOSTICS
LS100
10789-408PK
267.79
USD
10789-408
CompactDry™ LS, Listeria spp, Hardy Diagnostics
Microbiology Test Systems
Microorganism Tests
For the isolation, presumptive identification and enumeration of Listeria spp.
- Room temperature storage
- Self diffusing media
- Stackable
- Write-on area
- Easy to handle and pick up
- Simple to pick colonies for subculturing
- Easy read-out of results
CompactDry™ LS for the isolation, presumptive identification and enumeration of Listeria spp
This product is not intended to be used for the diagnosis of human disease.
Colonies of Listeria will turn blue due to the presence of chromogenic substances.