Medical Action Industries Biohazard Bag, Hardy Diagnostics
Supplier: Hardy Diagnostics
BB2305
10789-388EA 198.97 USD
10789-388
Medical Action Industries Biohazard Bag, Hardy Diagnostics
Bags Waste Disposal Bags
This 25×35", Biohazard bag is for the collection of biohazard waste.
  • Passed the 165 gm dart impact test
  • For collection of biohazard waste, not for autoclaving
Made of polyethylene, solid red with black biohazard warnings in English and Spanish, has the international biohazard symbol.
Certifications: Meets OSHA requirements.