VWR® PCR Workstations
Supplier: VWR International
VWRI732-2772
10783-132EA 3541.22 USD
10783-132
VWR® PCR Workstations
Fume Hoods and Enclosures PCR Workstation Enclosures
The VWR® PCR Workstation offers twin decontamination action by UV inactivation of airborne and surface-bound contaminants, and therefore represents an ideal environment for PCR sample preparation and other sensitive protocols.
  • Large working area for stress-free working
  • Can accommodate benchtop equipment
  • Function indicator for UV air recirculator tube
  • Displays operating time of UV tubes
  • Enables well-timed replacement of tubes for constant UV intensity
  • Removable shelves provide additional storage space at the rear panel
  • Four internal power outlets for
More Product Information
Order Now
SPECIFICATIONS
Working Area 72L×54D cm
Ext. Dimensions 75L×78H×62D cm