Cell-Chex Auto Controls, Streck
Supplier: STRECK LABORATORIES
200066 200069 200067 200068 200065 213479
89202-706PK 417.84 USD
89202-706 82030-700 82030-696 82030-698 87005-444 10783-062
Cell-Chex Auto Controls, Streck
Controls and Calibration Verification Materials
Cellular controls for evaluating the accuracy and precision of hematology instruments that measure blood cell counts in patient body fluid samples
The assay provides total RBC and WBC values. These multi-level controls are packaged in plastic vials with pierceable caps. Controls have an open-vial stability for 30 days and a closed-vial stability for 75 days when stored at 2 to 10 °C (36 to 50 °F). Compatible with Abbott®, Beckman Coulter®, and Sysmex® instruments.