Anti-GNB3 Chicken Polyclonal Antibody

Supplier: Biosensis
C-1520-100
10781-938EA 353.85 USD
10781-938
Anti-GNB3 Chicken Polyclonal Antibody
Antibodies
G-Protein beta3 (GNB3) is a guanine nucleotide-binding protein (G protein). G-proteins are involved as a modulator or transducer in various transmembrane signaling systems (Ref: SWISSPROT).

The antibody reacts specifically with GNB3 protein of human, mouse, and rat. Because of homology it is expects to react with other mammalian species as well. The antibody detects a prominent band ~38 kDa in western blot with extracts from human adipose tissue.
Application Information:
WB. Suggested dilution at 1:1,000 to 1:5,000. Biosensis recommends that the optimal working dilution should be determined by the end user.

Type: Primary
Antigen: G-Protein subunit beta-3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Human GNB3 amino acids 216-230.
Host: Chicken
Isotype: IgY
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR