Human IL-17E Standard ABTS ELISA Development Kit, PeproTech, Inc.
Supplier: Peprotech
900-K234
10777-894KT 160.21 USD
10777-894 10781-650
Human IL-17E Standard ABTS ELISA Development Kit, PeproTech, Inc.
Assays ELISAs
PeproTech's Human IL-17E ELISA development kit contains the key components required for the quantitative measurement of natural and/or recombinant human IL-17E in a sandwich ELISA format.
  • Easy-to-use
  • Optimized reagents for singleplex protein detection by solid phase sandwich enzyme-linked immunosorbent assay (ELISA)
Human IL-17E ELISA development kit contains the key components required for the quantitative measurement of natural and/or recombinant human IL-17E in a sandwich ELISA format. Using the ELISA protocol, the recommended microplates, reagents and solutions, the components supplied in this kit are sufficient to assay human IL-17E in approximately 1000 ELISA plate wells.