Murine sRANK Ligand Standard ABTS ELISA Development Kit, PeproTech
Supplier: Peprotech
900-K233
10777-892KT 173.57 USD
10777-892 10781-648
Murine sRANK Ligand Standard ABTS ELISA Development Kit, PeproTech
Assays ELISAs
PeproTech's Murine sRANKL ELISA development kit contains the key components required for the quantitative measurement of natural and/or recombinant murine sRANKL in a sandwich ELISA format.
  • Easy-to-use
  • Optimized reagents for singleplex protein detection by solid phase sandwich enzyme-linked immunosorbent assay (ELISA)
Murine sRANKL ELISA development kit contains the key components required for the quantitative measurement of natural and/or recombinant murine sRANKL in a sandwich ELISA format. Using the ELISA protocol, the recommended microplates, reagents and solutions, the components supplied in this kit are sufficient to assay murine sRANKL in approximately 1000 ELISA plate wells.