Human Recombinant NT-3 (from E. coli)
Supplier: Peprotech
450-03-2UG 450-03-10UG 450-03-100UG 450-03-250UG 450-03-500UG 450-03-1MG
10781-166EA 146.18 USD
10781-166 10781-168 10781-170 10781-172 10781-174 10781-176 10781-178
Human Recombinant NT-3 (from E. coli)
Proteins and Peptides
NT-3 is a neurotrophic factor structurally related to β-NGF, BDNF, and NT-4. These proteins belong to the cysteine-knot family of growth factors that assume stable dimeric structures. NT-3 is expressed by neurons of the central nervous systems, and can signal through the trk receptors. NT-3 promotes the growth and survival of nerve and glial cells. The amino acid sequences of human, murine and rat NT-3 are identical. Recombinant Human NT-3 is a noncovalently linked homodimer of two 13.6 kDa polypeptide monomers (240 total amino acid residues).
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.