Rat Recombinant IL-1 Alpha (from E. coli)
Supplier: Peprotech
400-01A-2UG 400-01A-10UG 400-01A-50UG 400-01A-100UG 400-01A-250UG 400-01A-500UG 400-01A-1MG
10780-842EA 132.81 USD
10780-842 10780-844 10780-846 10780-848 10780-850 10780-852 10780-854
Rat Recombinant IL-1 Alpha (from E. coli)
Proteins and Peptides
IL-1α is a non-secreted, proinflammatory cytokine produced in a variety of cells, including monocytes, tissue macrophages, keratinocytes, and other epithelial cells. Both IL-1α and IL-1β bind to the same receptor and have similar, if not identical, biological properties. These cytokines have a broad range of activities including the stimulation of thymocyte proliferation by inducing IL-2 release, B-cell maturation and proliferation, mitogenic FGF-like activity, and the release of prostaglandin and collagenase from synovial cells. However, whereas IL-1β is a secreted cytokine, IL-1α is predominantly a cell-associated cytokine. Recombinant Rat IL-1α is a 17.7 kDa protein containing 155 amino acid residues.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.