Human Cytokine
Supplier: Peprotech
AF-HHSC3
10778-868EA 4224.78 USD
10778-868
Human Cytokine
Proteins and Peptides
The Animal-Free Human Hematopoietic Stem Cell Expansion Cytokine Package contains the key components required for ex vivo human hematopoietic stem cells.
  • Animal-free
The package contains the following 4 vials:
100ug Animal-Free Human Flt3 Ligand
100ug Animal-Free Human SCF
100ug Animal-Free Human TPO
10ug Animal-Free Human IL-3
To request a CoA or MSDS for the components of the package, please Contact Us.
Manufactured using all Animal-Free reagents.
To request a CoA or MSDS for the components of the package, please Contact Us.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.