Mouse;Rat IL4/GM-CSF MDC
Supplier: Peprotech
MDC
10778-854EA 1606.12 USD
10778-854
Mouse;Rat IL4/GM-CSF MDC
Proteins and Peptides
The Murine Dendritic Cell Cytokine Package contains the key components required for the culturing of your mouse dendritic cells.
The package contains the following 2 vials:
100ug Murine IL-4
100ug Murine GM-CSF
To request a CoA or MSDS for the components of the package, please Contact Us.
To request a CoA or MSDS for the components of the package, please Contact Us.
Carrier free.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.