Human IL-4/GM-CSF HDC
Supplier: Peprotech
HDC
10778-852EA 1617.66 USD
10778-852
Human IL-4/GM-CSF HDC
Proteins and Peptides
The Human Dendritic Cell Cytokine Package contains the key components required for the culturing of your human dendritic cells.
The package contains the following 2 vials:
100ug Human IL-4
100ug Human GM-CSF
To request a CoA or MSDS for the components of the package, please Contact Us.
To request a CoA or MSDS for the components of the package, please Contact Us.
Carrier free.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.