Mouse Recombinant TPO (from E. coli)
Supplier: Peprotech
AF-315-14-2UG AF-315-14-10UG AF-315-14-50UG AF-315-14-100UG
10778-842EA 180.86 USD
10778-842 10778-844 10778-846 10778-848
Mouse Recombinant TPO (from E. coli)
Proteins and Peptides
TPO is a lineage-specific growth factor produced in the liver, kidney and skeletal muscle. It stimulates the proliferation and maturation of megakaryocytes, and promotes increased circulating levels of platelets in vivo . TPO signals through the c-mpl receptor, and acts as an important regulator of circulating platelets. Human and murine TPO exhibit cross-species reactivity. Recombinant Murine TPO is a fully biologically active 174 amino acid polypeptide (18.7 kDa), which contains the erythropoietin-like domain of the full length TPO protein.
  • Animal-free
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.