Murine MIP-1β (CCL4) Standard ABTS ELISA Development Kit, PeproTech
Supplier: Peprotech
900-K278
10777-718KT 577.11 USD
10777-718 10781-664
Murine MIP-1β (CCL4) Standard ABTS ELISA Development Kit, PeproTech
Assays ELISAs
PeproTech's Murine MIP-1β (CCL4) ELISA development kit contains the key components required for the quantitative measurement of natural and/or recombinant murine MIP-1β (CCL4) in a sandwich ELISA format.
  • Easy-to-use
  • Optimized reagents for singleplex protein detection by solid phase sandwich enzyme-linked immunosorbent assay (ELISA)
Murine MIP-1β (CCL4) ELISA development kit contains the key components required for the quantitative measurement of natural and/or recombinant murine MIP-1β (CCL4) in a sandwich ELISA format. Using the ELISA protocol, the recommended microplates, reagents and solutions, the components supplied in this kit are sufficient to assay murine MIP-1β (CCL4) in approximately 1000 ELISA plate wells.