Human MCP-2 (CCL8) Standard ABTS ELISA Development Kit, PeproTech
Supplier: Peprotech
900-K41
10777-682KT 577.11 USD
10777-682 10777-806
Human MCP-2 (CCL8) Standard ABTS ELISA Development Kit, PeproTech
Assays ELISAs
PeproTech's Human MCP-2 (CCL8) ELISA development kit contains the key components required for the quantitative measurement of natural and/or recombinant human MCP-2 (CCL8) in a sandwich ELISA format.
Human MCP-2 (CCL8) ELISA development kit contains the key components required for the quantitative measurement of natural and/or recombinant human MCP-2 (CCL8) in a sandwich ELISA format. Using the ELISA protocol, the recommended microplates, reagents and solutions, the components supplied in this kit are sufficient to assay human MCP-2 (CCL8) in approximately 1000 ELISA plate wells.