Anti-BMP-7 Mouse Monoclonal Antibody

Supplier: Peprotech
500-M123-500UG
10774-918EA 252.77 USD
10774-918
Anti-BMP-7 Mouse Monoclonal Antibody
Antibodies
Monoclonal AntibodyProduced in mice immunized with highly pure Recombinant Human BMP-7. Anti­Human BMP-7­specific antibody was purified from cell culture by Protein G affinity chromatography.

Applications tested can vary by lot number. See Product Data Sheet for more information.

Type: Primary
Antigen: BMP-7
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: Mouse IgG1k
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR