Anti-BMP-4 Mouse Monoclonal Antibody

Supplier: Peprotech
500-M121-500UG
10774-916EA 252.77 USD
10774-916
Anti-BMP-4 Mouse Monoclonal Antibody
Antibodies
Monoclonal AntibodyProduced in mice immunized with highly pure Recombinant Human BMP-4. Anti­Human BMP-4­specific antibody was purified from cell culture by Protein G affinity chromatography.

Applications tested can vary by lot number. See Product Data Sheet for more information.

Type: Primary
Antigen: BMP-4
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: Mouse IgG2bk
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR