Anti-beta-NGF Mouse Monoclonal Antibody

Supplier: Peprotech
500-M85-500UG
10774-906EA 252.77 USD
10774-906
Anti-beta-NGF Mouse Monoclonal Antibody
Antibodies
Monoclonal AntibodyProduced in mice immunized with highly pure Recombinant β-NGF. Anti­β-NGF­specific antibody was purified from cell culture by Protein G affinity chromatography.

Applications tested can vary by lot number. See Product Data Sheet for more information.

Type: Primary
Antigen: beta-NGF
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: Mouse IgG2bk
Reactivity: beta-NGF
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR