Anti-NT-4 Mouse Monoclonal Antibody

Supplier: Peprotech
500-M24-500UG
10774-850EA 252.77 USD
10774-850
Anti-NT-4 Mouse Monoclonal Antibody
Antibodies
Monoclonal AntibodyProduced in BALB/c x C57BL/6 F1-mice immunized with highly pure Recombinant Human NT-4. Anti­Human NT-4­specific antibody was purified from ascites fluid by Protein A affinity chromatography.

Applications tested can vary by lot number. See Product Data Sheet for more information.

Type: Primary
Antigen: NT-4
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: Mouse IgG2Ak
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR