Anti-IgG Mouse Monoclonal Antibody IL-8 (CXCL8)

Supplier: Peprotech
500-M08-500UG
10774-842EA 252.77 USD
10774-842
Anti-IgG Mouse Monoclonal Antibody IL-8 (CXCL8)
Antibodies
Produced in mice immunized with highly pure Recombinant Human IL-8 (CXCL8). Anti-Human IL-8 (CXCL8)-specific antibody was purified from cell culture by Protein G affinity chromatography.

Type: Primary
Antigen: IgG
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR