Anti-IL-4 Mouse Monoclonal Antibody

Supplier: Peprotech
500-M04-500UG
10774-836EA 272.21 USD
10774-836
Anti-IL-4 Mouse Monoclonal Antibody
Antibodies
Monoclonal AntibodyProduced in BALB/c x ICR F1-mice immunized with highly pure Recombinant Human IL-4. Anti­Human IL-4­specific antibody was purified from ascites fluid by Protein A affinity chromatography.

Applications tested can vary by lot number. See Product Data Sheet for more information.

Type: Primary
Antigen: IL-4
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: Mouse IgG1k
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR