Anti-IL-3 Mouse Monoclonal Antibody

Supplier: Peprotech
500-M03-500UG
10774-834EA 264.07 USD
10774-834
Anti-IL-3 Mouse Monoclonal Antibody
Antibodies
Monoclonal AntibodyProduced in mice immunized with highly pure Recombinant Human IL-3. Anti­Human IL-3­specific antibody was purified from tissue culture supernatant by Protein G affinity chromatography.

Applications tested can vary by lot number. See Product Data Sheet for more information.

Type: Primary
Antigen: IL-3
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: Mouse IgG1k
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR