Human Recombinant Chemerin (from E. coli)
Supplier: Peprotech
300-66-5UG 300-66-25UG 300-66-100UG 300-66-250UG 300-66-500UG 300-66-1MG
10774-110EA 132.81 USD
10774-110 10774-112 10774-114 10774-116 10774-118 10774-120
Human Recombinant Chemerin (from E. coli)
Proteins and Peptides
Chemerin is a secreted chemoattractant protein that can signal through the chemokine-like receptor-1 (CMKLR1). It is expressed in various tissues, including white adipose tissue, and circulates in blood as an inactive 143 amino acid precursor protein. Biologically active Chemerin is generated by proteolytic removal of C-terminal residues by several circulating proteases. Chemerin acts as a chemoattractant for cells expressing the CMKLR1 receptor, which includes certain dendritic cells, macrophages, and adipocytes. Recombinant Human Chemerin is a 15.6 kDa protein consisting of 135 amino acid residues.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.