Human Recombinant TECK (from E. coli)
Supplier: Peprotech
300-45-5UG 300-45-20UG 300-45-50UG 300-45-100UG 300-45-250UG 300-45-500UG 300-45-1MG
10773-992EA 140.15 USD
10773-992 10773-994 10773-996 10773-998 10774-000 10774-002 10774-004
Human Recombinant TECK (from E. coli)
Proteins and Peptides
TECK is a CC chemokine, specifically expressed by thymic stromal cells, and signals through the CCR9 receptor. TECK is chemotactic towards activated macrophages, thymocytes and dendritic cells. Recombinant Human TECK (CCL25) is a 14.2 kDa protein containing 127 amino acid residues, including the four conserved cysteine residues present in CC chemokines.
Ordering information: For research use only. Not for use in diagnostic or therapeutic procedures.