Glass Slide Thermal Transfer Label Kit
Supplier: Computype
1004781-1
10769-992KT 950.55 USD
10769-992
Glass Slide Thermal Transfer Label Kit
Labels
Glass Slide thermal transfer label kit is designed for use with glass slides exposed to chemical processing.
They are capable of holding up to direct immersion in xylene, DMSO, methanol, isopropyl alcohol, and staining. Labels are kitted with matched ribbon and label stock to ensure a solution durable for virtually any process and environment.