Pipet Helper®, Eppendorf
Supplier: Eppendorf
4423000010
10769-534EA 136.53 USD
10769-534
Pipet Helper®, Eppendorf
Pipettes Pipette Controllers
Pipet Helper's specially designed aspirating cone ensures secure seating for all measuring and volumetric pipettes (glass and plastic).
  • Drop wise dispensing and blow out without splashing
  • Drainage without any additional pressure delivers very reproducible results
  • Drip stop via hydrophobic membrane filter (3 µm)
  • Silicone pipette adapter guarantees a secure pipette fit
This is the perfect instrument for inexperienced users as its robust and intuitive design allows everybody to adjust the meniscus precisely
It is lightweight with reduced operating force. Easy disassembly, few parts and full autoclavability ease maintenance and cleaning. The aspirating bulb enables pipetting of volumes up to 100 ml at once. Single lever controls low force aspirating and dispensing. High control over the liquid movement due to very sensitive valve unit. It is mainly used for the preparation of a buffer for chromatography, fluid for fermentation, sample transfer within petrochemistry, generating a sample within water analytics, and for the preparation of assays.
In addition the valve unit is optimized so that liquids can be drawn up simply, without exerting pressure. The aspiration bulb provides rapid filling of the pipette. The aspiration and dispensing of liquids are controlled gently by the ergonomic lever. Additionally, a splash free blow-out is provided.