HyFlex® 11-318 Touch Screen-Capable Gloves, Uncoated, Ansell
Supplier: Ansell Healthcare
256366 256367 256368 256369 256370 256371
10769-420CS 2497.86 USD
10769-420 10769-494 10769-496 10769-498 10769-500 10769-502
HyFlex® 11-318 Touch Screen-Capable Gloves, Uncoated, Ansell
Gloves
Precision engineered to protect your hands and the product, the HyFlex® 11-318 are ultralight design gloves with medium cut performance.
  • Silicone-free
  • Superior tactile sensitivity
  • Ultimate flexibility
  • Low-linting
Featuring an 18 gauge knit with Dyneema® Diamond, Spandex, Nylon and Carbon to provide ultimate tactility and flexibility.
The ultralight fabric and tailored fit of the Hyflex 11-318 create a bare hand-like sensation of comfort and dexterity when performing tasks that demand high tactility. As the first-to-market glove to provide EN Level 3 cut protection in an 18 gauge style, the Hyflex 11-318 brings a new degree of dexterity to applications with cut risks. The new HyFlex 11-318 ensures the process/product quality because it is silicone-free and has antistatic properties.
The thin liner is uncoated, maintaining the hand at a comfortable temperature. Spandex reinforcement in the tension areas of the hand (knuckles and palm) enable a snug, anatomical fit on the hand. The HyFlex 11-318 is compatible with touch screens, so that workers do not need to take the glove off to handle electronic and touch screen devices, enabling higher efficiency in the workplace.
For use in packaging, quality inspection before painting, and final assembly of sharp parts.
Certifications: ANSI Cut Level 2. ANSI Abrasion Level 1.
Packaging: Packaged 12 pairs per polybag; 12 polybags per carton; 144 pairs per carton.