Apiezon AP101 Ultra High Vacuum Lubricating Grease, Silicone-Free, M&I Materials Ltd
Supplier: M & I Materials
AP101-00050
10769-150EA 292.92 USD
10769-150
Apiezon AP101 Ultra High Vacuum Lubricating Grease, Silicone-Free, M&I Materials Ltd
Lubricants
Apiezon AP101 is a specialist, anti-seize vacuum grease, containing PTFE, which confers superior anti-seize properties providing glass and metal surfaces with long-lasting lubrication and protection from fusing and corrosion.
  • Ideally suited for use in low to medium vacuum ranges having a vapour pressure of <10-5 Torr at 20 °C
  • Anti-seize properties, making it an effective protector and lubricant for metal surfaces which may be subject to seizure or corrosion
  • Critical lubricant
  • Insoluble in most solvents, resistant to water, alcohols, ketones and esters
  • Resists attack from aqueous acid and alkali solutions, alcoholic alkali solutions and corrosive gases
  • Suitable for use over a wide temperature range
  • Does not suffer from contamination problems associated with silicone based greases such as creep or carry over
  • Easy to clean and remove using hot water and an aqueous glassware detergent or hydrocarbon solvents