Right Angle Tee Joint to Tube Compression Adapter, Ace Glass Incorporated
Supplier: Ace Glass
12737-02 12737-04 12737-06 12737-08 12737-10 12737-12 12737-14 12737-50 12737-52 12737-54 12737-56 12737-58 12737-60 12737-62 12737-64 12737-16 12737-18 12737-20 12737-22 12737-24 12737-26 12737-28 12737-30 12737-32 12737-34 12737-36 12737-38 12737-40 12737-42 12737-44 12737-46 12737-48 12737-66 12737-68 12737-70 12737-72 12737-74
10767-498EA 60.15 USD
10767-498 10767-500 10767-502 10767-504 10767-506 10767-508 10767-510 10767-844 10767-846 10767-848 10767-850 10767-852 10767-854 10767-856 10767-858 10768-014 10768-016 10768-018 10768-020 10768-022 10768-024 10768-026 10768-028 10768-030 10768-032 10768-034 10768-036 10768-038 10768-040 10768-042 10768-044 10768-046 10768-048 10768-050 10768-052 10768-054 10768-056
Right Angle Tee Joint to Tube Compression Adapter, Ace Glass Incorporated
Adapters Adapters, Joints with Tube Connectors
These glass standard taper and spherical joint adapters feature right angle tee ground glass sidearms.
  • Glass [ST] or spherical joint
  • Right angle tee sidearms
The sidearms are used to make tube compression fitting connections to a wide variety of flexible and rigid tubing using PTFE fittings.