Right Angle Tube Compression Adapter, with Drip Tip, Ace Glass Incorporated
Supplier: Ace Glass
12722-01 12722-03 12722-17 12722-19 12722-21 12722-23 12722-07 12722-09 12722-11 12722-13 12722-15 12722-25 12722-27
10767-460EA 76.07 USD
10767-460 10767-462 10767-804 10767-806 10767-808 10767-810 10767-966 10767-968 10767-970 10767-972 10767-974 10767-976 10767-978
Right Angle Tube Compression Adapter, with Drip Tip, Ace Glass Incorporated
Adapters Adapters, Joints with Tube Connectors
The glass standard taper joint adapters feature a drip tip and a right angle ground glass tube sidearm.
  • Glass [ST] joint
  • Drip tip
  • Right angle sidearm
The sidearm is used to make tube compression fitting connections to a wide variety of flexible and rigid tubing using PTFE fittings.