Voltage Controller, Glas-Col Powrtrol, Ace Glass Incorporated
Supplier: Ace Glass
12089-30 12089-40
10767-338EA 678.65 USD
10767-338 10767-340
Voltage Controller, Glas-Col Powrtrol, Ace Glass Incorporated
Controllers Utility Controllers
Solid-state proportional-voltage power control that provides precise manual control of heating mantles, tapes, cords, and other resistive loads.
  • The output range is conveniently adjustable from 5-100% of rated voltage
  • One year manufacturer's limited warranty.
Foxtip gray and charcoal, low profile lab bench cabinet with clamp.
Ordering information: 120 V/60 Hz/1200 W and 240 V/50 Hz/2400 W models available.