GL 45 Connection System, Ace Glass Incorporated
Supplier: Ace Glass
7634-01 7634-02
10767-308PK 284.34 USD
10767-308 10767-310 10767-312
GL 45 Connection System, Ace Glass Incorporated
Closures Bottle Closures
GL45 cap, female syringe luer slip, syringe filter connector for Duran connection system
Possible applications included the transfer of sterile media from one container to another using a perstaltic pump.
Cap has swiveling, dual propylene hose connectors for use with soft elastic tubing that has an ID of 6-9 mm. The cap's center rotates freely, allowing the bottle cap to be unscrewed without the need to disconnect the tubing. Polypropylene cap is temperature resistant to 140°C.