TransIT®-QR Delivery Solution and Starter Kit, Mirus Bio
Supplier: Mirus Bio
MIR5240
10767-006EA 246.1 USD
10767-006
TransIT®-QR Delivery Solution and Starter Kit, Mirus Bio
Transfection Reagents
A nontoxic, "Quick Recovery" in vivo delivery solution that delivers DNA or siRNA to the livers of laboratory mice using hydrodynamic tail vein injection.
  • Naked nucleic acid delivery
  • Versatile platform
  • Low toxicity
Each TransIT®-QR Starter Kit contains the TransIT®-QR Delivery Solution, 10 syringes, needles and alcohol swabs, and a conical plastic tube to restrain the mouse while tail vein injection is performed. The TransIT®-QR Delivery Solution is also sold separately as indicated above.