Puromycin dihydrochloride solution

Supplier: Mirus Bio

Synonyms: (S)-2-Amino-N-((2S,3S,4R,5R)-5-(6-(Dimethylamino)-9H-purin-9-yl)-4-hydroxy-2-(hydroxymethyl)tetrahydrofuran-3-yl)-3-(4-methoxyphenyl)propanamide dihydrochloride

MIR5940
10766-886EA 382.81 USD
10766-886
Puromycin dihydrochloride solution
Puromycin dihydrochloride

Puromycin antibiotic ensures effective positive selection of cells expressing the puromycin-N-acetyltransferase (pac) gene.


  • recommended working concentration range is 0.5 to 10 µg/mL in mammalian cells
  • Concentration of 10 mg/mL in DI Water
  • Store at –20 °C


Different cell types and cell culture conditions may require different concentrations of selection antibiotic.

Formula: C₂₂H₂₉N₇O₅·2HCl
MW: 544.44 g/mol
MDL Number: MFCD00012691
CAS Number: 58-58-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR