G418 disulfate solution

Supplier: Mirus Bio

MIR5920
10766-882EA 122.05 USD
10766-882
G418 disulfate solution
G418 disulphate

G418 antibiotic solution is used for stable cell line generation, it ensures effective positive selection for cells expressing the neomycin resistance (neo) gene. Different cell types and cell culture conditions may require different concentrations of selection antibiotic.


  • Recommended working concentration range is 0.1 to 2.0 mg/mL in mammalian cells
  • Store at 4 °C
  • To be protected from light


Concentration: 50 mg/mL G418 Sulfate, sterile filtered in DI water.

Formula: C₂₀H₄₄N₄O₁₈S₂
MDL Number: MFCD00058314
CAS Number: 108321-42-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR