Stainless Steel Specimen Chuck Stand, Springside Scientific
Supplier: SPRINGSIDE SCIENTIFIC, LLC.
SDST001
10764-046EA 100.05 USD
10764-046
Stainless Steel Specimen Chuck Stand, Springside Scientific
Specimen Mounting Supplies
This stainless steel stand is used to store chucks both outside and inside the cooling chamber.
This small metal stand fits five specimen chucks up to 55 mm in diameter. Specimen chucks are not included. High quality stainless steel stand is perfect for in-chamber and can be used to expand the internal chuck capacity of the cryostat.