Hyaline Synthetic Low Temperature Cryostat Lubrication Kit, Springside Scientific
Supplier: SPRINGSIDE SCIENTIFIC, LLC.
SPCLK04
10764-044KT 190.93 USD
10764-044
Hyaline Synthetic Low Temperature Cryostat Lubrication Kit, Springside Scientific
Lubricants
This product is a quality alternative to Leica and is made from the highest quality, low-temperature synthetic lubricants.
  • Special low temperature food grade synthetic lubricants
  • Tested to temperatures down to –70 °C
  • Lubricants are safe and odorless
  • Light oil perfect for for cross bearing assemblies and any shaft roller bearings
Low temperature synthetic grease is perfect for mating surfaces of the cutting block, cutting head moving parts or the internal sliding mechanisms within the microtome.
Delivery information: Kit includes 2 x 1 oz. ultrapure cryostat oil and 2 x 2 oz. brush-on synthetic cryostat grease.