Anti-TCF1 Mouse Monoclonal Antibody [clone: 7F11A10]

Supplier: Biolegend
655202
10763-526EA 327.55 USD
10763-526
Anti-TCF1 Mouse Monoclonal Antibody [clone: 7F11A10]
Antibodies
Purified anti-TCF1 (TCF7) [7F11A10]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: WB, IF; Size: 100 μg

Each lot of this antibody is quality control tested by Western blotting. For Western blotting, the suggested use of this reagent is 0.1 - 0.5 μg per ml. For immunofluorescence microscopy, a concentration range of 1-5 μg/ml is recommended. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: TCF1
Clonality: Monoclonal
Clone: 7F11A10
Conjugation:
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR