Anti-SWAP70 Mouse Monoclonal Antibody [clone: 9D9A19]

Supplier: Biolegend
654702
10763-512EA 298.3 USD
10763-512
Anti-SWAP70 Mouse Monoclonal Antibody [clone: 9D9A19]
Antibodies
Purified anti-SWAP70 [9D9A19]; Isotype: Mouse IgG1, κ; Reactivity: Human; Apps: WB, IP; Size: 100 μg

Each lot of this antibody is quality control tested by Western blotting. For Western blotting, the suggested use of this reagent is 0.1-0.5 μg per ml. For immunoprecipitation, the suggested use of this reagent is 10-30 μg per ml. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: SWAP70
Clonality: Monoclonal
Clone: 9D9A19
Conjugation:
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR