Anti-CD63 Mouse Monoclonal Antibody (PE (Phycoerythrin)/Dazzle 594) [clone: H5C6]

Supplier: Biolegend
353021 353022
10759-674EA 179 USD
10759-674 10759-676
Anti-CD63 Mouse Monoclonal Antibody (PE (Phycoerythrin)/Dazzle 594) [clone: H5C6]
Antibodies
PE/Dazzle™ 594 anti-human CD63 [H5C6]; Isotype: Mouse IgG1, κ; Reactivity: Human, African Green, Baboon, Cynomolgus, Rhesus; Apps: FC; Size: 100 tests

Each lot of this antibody is quality control tested by immunofluorescent staining with flow cytometric analysis. For flow cytometric staining, the suggested use of this reagent is 5 μl per million cells or 5 μl per 100 μl of whole blood. It is recommended that the reagent be titrated for optimal performance for each application.

Type: Primary
Antigen: CD63
Clonality: Monoclonal
Clone: H5C6
Conjugation: PE (Phycoerythrin)/Dazzle 594
Epitope:
Host: Mouse
Isotype: Mouse IgG1, κ
Reactivity: Human;African Green;Baboon;Cynomolgus;Rhesus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR